missing translation for 'onlineSavingsMsg'
Learn More

Palladium Silver foil, 0.025mm (0.001in) thick, 99.9% (metals basis excluding Pt)

Catalog No. AA42187FF
Change view
Click to view available options
Quantity:
25 x 25 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42187FF 25 x 25 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA42187FF Supplier Thermo Scientific Chemicals Supplier No. 42187FF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pd:Ag; 75:25 wt%

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

Molecular Formula Pd:Ag; 75:25 wt%
MDL Number MFCD02091738

Specifications

Packaging Tube
Quantity 25 x 25 mm
Percent Purity 99.9%
Chemical Name or Material Palladium Silver foil
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only