missing translation for 'onlineSavingsMsg'
Learn More

Palladium, Powdered 99+, MilliporeSigma™

Catalog No. p-4939468
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1192250001 1 g Glass bottle
M1192250005 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1192250001 Supplier Technidata Supplier No. 1.19225.0001
Only null left

Chemical Identifiers

CAS 5-3-7440
Molecular Formula Pd
Molecular Weight (g/mol) 106.42
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
Synonym black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
PubChem CID 23938
ChEBI CHEBI:33363
IUPAC Name palladium
SMILES [Pd]

Specifications

Appearance Gray powder
Quantity 1 g
Odor Odorless
Packaging Glass bottle
Form Powder
Formula Weight 106.4
Chemical Name or Material Palladium