missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) Trifluoroacetate 98.0+%, TCI America™

Catalog No. p-7135298
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18701G 1 g
P18705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P18701G Supplier TCI America Supplier No. P18701G
Only null left

Chemical Identifiers

CAS 42196-31-6
Molecular Formula C4F6O4Pd
Molecular Weight (g/mol) 332.45
MDL Number MFCD00013204
InChI Key PBDBXAQKXCXZCJ-UHFFFAOYSA-L
Synonym palladium ii trifluoroacetate, palladium ii 2,2,2-trifluoroacetate, palladium 2+ ion ditrifluoroacetate, palladium trifluoroacetate, palladium 2+ ditrifluoroacetate, trifluoroacetic acid palladium ii salt, bis 2,2,2-trifluoroacetoxy palladium, acmc-209jo7, ksc235m3d
PubChem CID 2733347
IUPAC Name palladium(2+) ditrifluoroacetate
SMILES [Pd++].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F

Specifications

Melting Point 220°C
Quantity 1 g
Formula Weight 332.45
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Palladium(II) Trifluoroacetate