Learn More
Palladium(II) nitrate hydrate
CAS: 207596-32-5 | N2O6Pd | 230.43 g/mol
$343.57 - $7516.08
Chemical Identifiers
| CAS | 207596-32-5 |
|---|---|
| Molecular Formula | N2O6Pd |
| Molecular Weight (g/mol) | 230.43 |
| MDL Number | MFCD00149818 |
| InChI Key | GPNDARIEYHPYAY-UHFFFAOYSA-N |
| Synonym | palladium ii nitrate hydrate, palladium nitrate hydrate, palladium 2+ hydrate dinitrate, acmc-1bnae, palladous nitrate hydrate, palladium dinitrate hydrate, 2no3.pd.h2o, ksc492s0r, palladium ii nitrate,hydrate, palladium,diaquabis nitrato-o-9ci |
| PubChem CID | 16211503 |
| SMILES | [Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC389910010
|
Thermo Scientific Chemicals
389910010 |
1 g | Glass bottle |
Each for $343.57
|
|
||||
|
AC389910250
|
Thermo Scientific Chemicals
389910250 |
25 g | Glass bottle |
Each for $7,516.08
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 207596-32-5 | |
| 230.43 | |
| GPNDARIEYHPYAY-UHFFFAOYSA-N | |
| 16211503 |
| N2O6Pd | |
| MFCD00149818 | |
| palladium ii nitrate hydrate, palladium nitrate hydrate, palladium 2+ hydrate dinitrate, acmc-1bnae, palladous nitrate hydrate, palladium dinitrate hydrate, 2no3.pd.h2o, ksc492s0r, palladium ii nitrate,hydrate, palladium,diaquabis nitrato-o-9ci | |
| [Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
| Black-Brown | |
| Crystals | |
| N2O6Pd | |
| MFCD00149818 | |
| GPNDARIEYHPYAY-UHFFFAOYSA-N | |
| palladium(2+) dinitrate | |
| 16211503 | |
| Glass bottle |
| 207596-32-5,10102-05-3 | |
| 1 g | |
| Pd(NO3)2·xH2O | |
| palladium ii nitrate hydrate, palladium nitrate hydrate, palladium 2+ hydrate dinitrate, acmc-1bnae, palladous nitrate hydrate, palladium dinitrate hydrate, 2no3.pd.h2o, ksc492s0r, palladium ii nitrate,hydrate, palladium,diaquabis nitrato-o-9ci | |
| [Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 230.43 | |
| 230.43 | |
| Palladium(II) nitrate hydrate |
Safety and Handling
GHS H Statement
May intensify fire; oxidizer.
May be corrosive to metals.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
RUO – Research Use Only