missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) Chloride 98.0+%, TCI America™

Catalog No. p-7135157
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14891G 1 g
P14895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P14891G Supplier TCI America Supplier No. P14891G
Only null left

Chemical Identifiers

CAS 7647-10-1
Molecular Formula Cl2Pd
Molecular Weight (g/mol) 177.32
MDL Number MFCD00003558
InChI Key PIBWKRNGBLPSSY-UHFFFAOYSA-L
Synonym palladium chloride, palladium ii chloride, palladium dichloride, palladous chloride, pdcl2, palladium 2+ chloride, palladium chloride pdcl2, enplate activator 440
PubChem CID 24290
ChEBI CHEBI:53434
IUPAC Name palladium(2+) dichloride
SMILES [Cl-].[Cl-].[Pd++]

Specifications

Melting Point 680°C
Physical Form Crystalline Powder
Quantity 1 g
Formula Weight 177.32
Percent Purity ≥98.0% (T)
Chemical Name or Material Palladium(II) Chloride
EINECSNumber (1)-0253
RTECSNumber RT3500000
TSCA Yes