missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium(II) Chloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 680°C |
| Physical Form | Crystalline Powder |
| Quantity | 1 g |
| Formula Weight | 177.32 |
| Percent Purity | ≥98.0% (T) |
| Chemical Name or Material | Palladium(II) Chloride |
Safety and Handling
| EINECSNumber | (1)-0253 |
| RTECSNumber | RT3500000 |
| TSCA | Yes |