missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) chloride, 59% Pd

Catalog No. p-4509774
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC195200010 1 g Glass bottle
AC195200050 5 g Glass bottle
AC195200250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC195200010 Supplier Thermo Scientific Chemicals Supplier No. 195200010
Only null left

CAS: 7647-10-1 | Cl2Pd | 177.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7647-10-1
Molecular Formula Cl2Pd
Molecular Weight (g/mol) 177.32
MDL Number MFCD00003558
InChI Key PIBWKRNGBLPSSY-UHFFFAOYSA-L
Synonym palladium chloride, palladium ii chloride, palladium dichloride, palladous chloride, pdcl2, palladium 2+ chloride, palladium chloride pdcl2, enplate activator 440
PubChem CID 24290
ChEBI CHEBI:53434
SMILES [Cl-].[Cl-].[Pd++]

Specifications

Melting Point 678.0°C to 680.0°C
Color Red to Brown
Physical Form Powder
Quantity 1 g
Linear Formula PdCl2
Fieser 01,782; 02,303; 04,369; 05,500; 06,447; 07,277; 08,384; 09,352; 10,300; 11,393; 12,371; 13,9; 15,248; 16,268
Merck Index 15,7091
Solubility Information Solubility in water: 31.99g/L. Other solubilities: soluble in hydrochloric acid,hydrobromic acid,soluble in alcohol and acetone
IUPAC Name palladium(2+) dichloride
Formula Weight 177.31
Percent Purity 59 to 60% (Pd)
Packaging Glass bottle
Chemical Name or Material Palladium(II) chloride
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May be corrosive to metals. May cause an allergic skin reaction. Causes serious eye damage.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Absorb spillage to prevent material damage. Keep only in original container.

RUO – Research Use Only