missing translation for 'onlineSavingsMsg'
Learn More

Palladium Hydroxide (contains Pd, PdO) on Carbon (wetted with ca. 50% Water), TCI America™

Catalog No. p-7135167
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P152810G 10 g
P152850G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P152810G Supplier TCI America Supplier No. P152810G
Only null left

Chemical Identifiers

CAS 12135-22-7
Molecular Formula H2O2Pd
Molecular Weight (g/mol) 140.43
MDL Number MFCD00064599
InChI Key NXJCBFBQEVOTOW-UHFFFAOYSA-L
Synonym palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon
PubChem CID 9942167
IUPAC Name palladium(2+) dihydroxide
SMILES [OH-].[OH-].[Pd++]

Specifications

Color Black
Physical Form Crystalline Powder
Quantity 10 g
Formula Weight 140.43
Chemical Name or Material Palladium Hydroxide (contains Pd, PdO) on Carbon (wetted with ca. 50% Water)
EINECSNumber (1)-1141
TSCA Yes