Learn More
Strem Chemicals Inc CAS# 1334385-47-5. 1g. Chloro(2-dicyclohexylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl)[2-(2-aminoethyl)phenyl] palladium(II) methyl-t-butylether adduct, min. 98% [XPhos Palladacycle Gen. 1]. MFCD11045094 DFS Item
Supplier:  Strem Chemicals Inc 4602681
CAS# 1334385-47-5. 1g. Chloro(2-dicyclohexylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl)[2-(2-aminoethyl)phenyl] palladium(II) methyl-t-butylether adduct, min. 98% [XPhos Palladacycle Gen. 1]. MFCD11045094. Molecular Weight: 738.76. Molecular Formula: C41H59ClNPPd. Color/form: white pwdr. Strem# 46-0268. http://www.strem.com/catalog/v/46-0268/
Catalog No. 5090112306
Only null left
Explore available promotions