missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium, 5% on carbon powder, Type 490
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 10 g |
| UN Number | UN1325 |
| Chemical Name or Material | Palladium, 5% on carbon powder, Type 490 |
Safety and Handling
| Hazard Category | H228 |
| Hazard Statement | GHS H Statement H228 Flammable solid. |
| Precautionary Statement | P210-P240-P241-P280-P370+P378q |
| DOTInformation | Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S. |
| EINECSNumber | 231-115-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only