missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium 5% on Alumina, TCI America™
Specifications
Specifications
| Color | Black |
| Quantity | 5 g |
| Formula Weight | 106.42 |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Palladium 5% on Alumina |
Safety and Handling
| RTECSNumber | RT3480500 |
| TSCA | Yes |