missing translation for 'onlineSavingsMsg'
Learn More

Palladium 10% on Carbon (wetted with ca. 55% Water), TCI America™

Catalog No. P17855G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17855G 5 g
P178525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P17855G Supplier TCI America Supplier No. P17855G
Only null left

Chemical Identifiers

CAS 5-3-7440
Molecular Formula Pd
Molecular Weight (g/mol) 106.42
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
Synonym black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
PubChem CID 23938
ChEBI CHEBI:33363
IUPAC Name palladium
SMILES [Pd]

Specifications

Color Black
Quantity 5 g
Formula Weight 106.42
Physical Form Crystalline Powder
Chemical Name or Material Palladium 10% on Carbon (wetted with ca. 55% Water) [Useful catalyst for coupling reaction, etc.]
RTECSNumber RT3480500
TSCA Yes