missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium 10% on Carbon (wetted with ca. 55% Water), TCI America™
Specifications
Specifications
| Color | Black |
| Quantity | 25 g |
| Form | Powder |
| Formula Weight | 106.42 |
| Chemical Name or Material | Palladium 10% on Carbon (wetted with ca. 55% Water) |
Safety and Handling
| RTECSNumber | RT3480500 |
| TSCA | Yes |