Learn More
SignalChem Biotech PAK1, Active 500ug
Catalog No. 502573946
Encompass
Recombinant full-length human PAK1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 502573946 Supplier SignalChem Biotech Supplier No. P02B10BG500
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant full-length human PAK1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.