Learn More
Fisherbrand™ Painted Steel Vacuum Pump Cart
Vacuum Pump Cabinets are standing height, 35.1 in. tall. They have sound deadening material included. Pump cart is sold separately. Standard color is Fisherbrand™ White.
$1427.55 - $2032.41
Specifications
Color White
Material Steel (18 ga.)
View More Specs
Products 3
Catalog Number Mfr. No. Dimensions (L x W x H) Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W x H) Price Quantity  
15050071
View Documents
N/A 16 x 19 x 5.7 in.
Each for $1,427.55
Only null left
15050072
View Documents
N/A 16 x 31 x 5.7 in.
Each for $1,488.03
Only null left
15050073
View Documents
N/A 16 x 43 x 5.7 in.
Each for $2,032.41
Only null left
Description
Description
  • Vacuum Pump Cart
  • Fisherbrand White is a urethane powder coating that provides reliable chemical and abrasion resistance
  • Locking swivel casters for ease of use to access vacuum pumps within cabinets
Specifications
Specifications
White
Steel (18 ga.)
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'