missing translation for 'onlineSavingsMsg'
Learn More

Paeonolide 97.0+%, TCI America™

Catalog No. P187910MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P187910MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P187910MG Supplier TCI America Supplier No. P187910MG
Only null left

Chemical Identifiers

CAS 72520-92-4
Molecular Formula C20H28O12
Molecular Weight (g/mol) 460.432
InChI Key IDZZECHGWAZTIB-NYBIBFQCSA-N
Synonym 2-Acetyl-5-methoxyphenyl 6-O-(alpha-L-Arabinopyranosyl)-beta-D-glucopyranoside
PubChem CID 442923
ChEBI CHEBI:7891
IUPAC Name 1-[4-methoxy-2-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2S,3R,4S,5S)-3,4,5-trihydroxyoxan-2-yl]oxymethyl]oxan-2-yl]oxyphenyl]ethanone
SMILES CC(=O)C1=C(C=C(C=C1)OC)OC2C(C(C(C(O2)COC3C(C(C(CO3)O)O)O)O)O)O

Specifications

Melting Point 197°C
Color White
Quantity 10 mg
Formula Weight 460.43
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Paeonolide
TSCA No
Recommended Storage Refrigerator