missing translation for 'onlineSavingsMsg'
Learn More

Paeoniflorin 97.0+%, TCI America™

Catalog No. p1876100mg
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mg
Catalog No. Quantity
P1876100MG 100 mg
1 options

Catalog No. P1876100MG

Supplier: TCI America P1876100MG

Only null left

Chemical Identifiers

CAS 23180-57-6
Molecular Formula C23H28O11
Molecular Weight (g/mol) 480.47
MDL Number MFCD00869331,MFCD00869331
InChI Key YKRGDOXKVOZESV-WRJNSLSBSA-N
PubChem CID 131674882
IUPAC Name [(1R,2S,3R,5R,6R,8S)-6-hydroxy-8-methyl-3-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-9,10-dioxatetracyclo[4.3.1.0²,⁵.0³,⁸]decan-2-yl]methyl benzoate
SMILES C[C@@]12C[C@@]3(O)O[C@@H](O1)[C@]1(COC(=O)C4=CC=CC=C4)[C@H]3C[C@]21O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

Specifications

Color White
Quantity 100 mg
Formula Weight 480.47
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Paeoniflorin
RTECSNumber RT1200000
TSCA No
Recommended Storage Refrigerator