missing translation for 'onlineSavingsMsg'
Learn More

Padimate O Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013535
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013535 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013535 Supplier Merck Emd Millipore Supplier No. PHR135510G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 21245-02-3
Density 0.995 g/mL (at 25°C)
Flash Point 113°C - closed cup
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Physical Form Neat
Linear Formula (CH3)2 NC6H4 CO2 CH2 CH(C2H5)(CH2)3 CH3
MDL Number MFCD00017526
Boiling Point 325°C
Synonym 2-Ethylhexyl 4-(dimethylamino)benzoate
Type Certified Reference Material
Quantity 10 g
Formula Weight 277.40
Grade Certified Reference Material
Show More Show Less