missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PADI4 Antibody Blocking Peptide

Catalog No. NB398494
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398494 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398494 Supplier Novus Biologicals™ Supplier No. NBP197309PEP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Blocking Peptide

Specifications

Gene ID (Entrez) 23569
Concentration 1 mg/mL
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.
For Use With (Application) This peptide is useful as a blocking peptide for NBP1-97309. For further blocking peptide related protocol, click here .
Gene Alias HL-60 PAD, PADI5PADI-H protein, PADPAD4, PDI4, PDI5EC 3.5.3.15, peptidyl arginine deiminase, type IV, peptidyl arginine deiminase, type V, Peptidylarginine deiminase IV, Protein-arginine deiminase type IV, protein-arginine deiminase type-4
Gene Symbol PADI4
Quantity 0.1 mg
Research Category Lipid and Metabolism
Storage Buffer Peptide dissolved in dH20. Contains no BSA.