missing translation for 'onlineSavingsMsg'
Learn More

Paclobutrazol 98.0+%, TCI America™

Catalog No. p-7135500
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22991G 1 g
P22995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22991G Supplier TCI America Supplier No. P22991G
Only null left

Chemical Identifiers

CAS 76738-62-0
Molecular Formula C15H20ClN3O
Molecular Weight (g/mol) 293.795
MDL Number MFCD01678673
InChI Key RMOGWMIKYWRTKW-LSLKUGRBSA-N
Synonym paclobutrazol, 2s-1-4-chlorophenyl-4,4-dimethyl-2-1,2,4-triazol-1-yl-3-pentanol, 2s-1-4-chlorophenyl-4,4-dimethyl-2-1,2,4-triazol-1-yl pentan-3-ol
PubChem CID 53297383
IUPAC Name (2S)-1-(4-chlorophenyl)-4,4-dimethyl-2-(1,2,4-triazol-1-yl)pentan-3-ol
SMILES CC(C)(C)C(C(CC1=CC=C(C=C1)Cl)N2C=NC=N2)O

Specifications

Melting Point 164°C
Color White
Quantity 1 g
UN Number 1325
Formula Weight 293.80
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Paclobutrazol
RTECSNumber XZ4803300
TSCA No