Learn More
Paclitaxel, 99.5+%
Paclitaxel (CAS # 33069-62-4) is a microtubule inhibitor produced from the extract of the Pacific yew tree. | CAS: 33069-62-4 | C47H51NO14 | 853.92 g/mol
Supplier: Thermo Scientific Chemicals J6273403
| Quantity | 1 g |
|---|
Description
Promotes assembly and inhibits disassembly of microtubules (anti-cancer agent)Paclitaxel is widely used in chemotherapeutic compound. It is used to treat certain types of cancer such as ovarian cancer, breast cancer, lung cancer and pancreatic cancer among others. It interferes with the growth of cancer cells and slows down their growth and spread in the body. It is also used to treat AIDS-related Kaposi's sarcoma.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Paclitaxel is a tetracyclic diterpenoid isolated from the Pacific yew, Taxus brevifolia
- Paclitaxel acts as an antineoplastic agent by binding tubulin and preventing cell division
- Paclitaxel may also bind Bcl-2 and lead to disinhibition of apoptosis
Application
- Paclitaxel is used in the study of lung, breast, and ovarian cancers; it may be useful in the research of other cancer types
- It is suitable for cell culture, biochemistry, and molecular biology applications
Chemical Identifiers
| 33069-62-4 | |
| 853.92 | |
| RCINICONZNJXQF-VAZQATRQSA-N | |
| 133640187 | |
| CC(=O)O[C@@H]1C2=C(C)[C@H](C[C@@](O)([C@@H](OC(=O)C3=CC=CC=C3)C3[C@@]4(CO[C@@H]4C[C@H](O)[C@@]3(C)C1=O)OC(C)=O)C2(C)C)OC(=O)[C@H](O)[C@@H](NC(=O)C1=CC=CC=C1)C1=CC=CC=C1 |
| C47H51NO14 | |
| MFCD00869953 | |
| paclitaxel, taxol, taxol a, abraxane, paxene, paxceed, plaxicel, yewtaxan, onxol, ebetaxel | |
| (1S,2S,4S,7R,9S,10S,12R,15S)-4,12-bis(acetyloxy)-1,9-dihydroxy-15-{[(2R,3S)-2-hydroxy-3-phenyl-3-(phenylformamido)propanoyl]oxy}-10,14,17,17-tetramethyl-11-oxo-6-oxatetracyclo[11.3.1.0³,¹⁰.0⁴,⁷]heptadec-13-en-2-yl benzoate |
Specifications
| Paclitaxel | |
| 213°C (decomposition) | |
| C47H51NO14 | |
| MFCD00869953 | |
| 14,6982 | |
| Soluble in DMSO (200mg/ml) or ethanol (40mg/ml); Soluble in dimethylsulfoxide,methanol,ethanol,acetonitrile,cremophor and dimehtylsulfamide; Sparingly soluble in aqueous buffer. | |
| CC(=O)O[C@@H]1C2=C(C)[C@H](C[C@@](O)([C@@H](OC(=O)C3=CC=CC=C3)C3[C@@]4(CO[C@@H]4C[C@H](O)[C@@]3(C)C1=O)OC(C)=O)C2(C)C)OC(=O)[C@H](O)[C@@H](NC(=O)C1=CC=CC=C1)C1=CC=CC=C1 | |
| 853.92 | |
| 853.91 | |
| Crystalline Solid |
| 33069-62-4 | |
| White | |
| 1 g | |
| 1420457 | |
| paclitaxel, taxol, taxol a, abraxane, paxene, paxceed, plaxicel, yewtaxan, onxol, ebetaxel | |
| RCINICONZNJXQF-VAZQATRQSA-N | |
| (1S,2S,4S,7R,9S,10S,12R,15S)-4,12-bis(acetyloxy)-1,9-dihydroxy-15-{[(2R,3S)-2-hydroxy-3-phenyl-3-(phenylformamido)propanoyl]oxy}-10,14,17,17-tetramethyl-11-oxo-6-oxatetracyclo[11.3.1.0³,¹⁰.0⁴,⁷]heptadec-13-en-2-yl benzoate | |
| 133640187 | |
| ≥99.5% |
Safety and Handling
GHS H Statement
H334-H341-H361-H318-H302-H312-H332-H335-H315-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.
P201-P202-P261-P264b-P270-P271-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P342+P311-P363-P501c
H302+H312+H332-H318-H334-H335-H361
RTECSNumber : DA8340700
TSCA : No
Recommended Storage : Store at -20°C