missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Paclitaxel, 99+%

Catalog No. AC328420250
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC328420250 25 mg
AC328420010 1 mg
AC328420050 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC328420250 Supplier Thermo Scientific Chemicals Supplier No. 328420250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Paclitaxel, also known as Taxol, CAS # 33069-62-4 is a chemotherapeutic drug that acts through stabilizing microtubule assembly and arresting cell cycle progression. | CAS: 33069-62-4 | C47H51NO14 | 853.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Paclitaxel is a naturally ocurring compound originally isolated from the tree T. brevifolia
  • Paclitaxel binds tubulin and stabilizes microtubule formation, preventing disassembly
  • Paclitaxel is used as a drug to arrest cells at the mitotic checkpoint and block cell cycle progression, thus inducing death of cancer cells

Applications

  • Paclitaxel can be used in preclinical laboratory research on diseases such as cancer, heart disease, inflammatory disorders, and hepatic fibrosis
  • Paclitaxel can be used as a tool in cell biology to study mitotic checkpoint arrest conditions and downstream signaling cascades

Chemical Identifiers

CAS 33069-62-4
Molecular Formula C47H51NO14
Molecular Weight (g/mol) 853.92
MDL Number MFCD00869953
InChI Key RCINICONZNJXQF-VAZQATRQSA-N
Synonym paclitaxel, taxol, taxol a, abraxane, paxene, paxceed, plaxicel, yewtaxan, onxol, ebetaxel
PubChem CID 133640187
IUPAC Name (1S,2S,4S,7R,9S,10S,12R,15S)-4,12-bis(acetyloxy)-1,9-dihydroxy-15-{[(2R,3S)-2-hydroxy-3-phenyl-3-(phenylformamido)propanoyl]oxy}-10,14,17,17-tetramethyl-11-oxo-6-oxatetracyclo[11.3.1.0³,¹⁰.0⁴,⁷]heptadec-13-en-2-yl benzoate
SMILES CC(=O)O[C@@H]1C2=C(C)[C@H](C[C@@](O)([C@@H](OC(=O)C3=CC=CC=C3)C3[C@@]4(CO[C@@H]4C[C@H](O)[C@@]3(C)C1=O)OC(C)=O)C2(C)C)OC(=O)[C@H](O)[C@@H](NC(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 213.0°C to 216.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Quantity 25 mg
Merck Index 15, 7081
Solubility Information Solubility in water: sparingly soluble. Other solubilities: soluble in methanol,dmso,acetonitrile
Formula Weight 853.9
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material Paclitaxel, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Suspected of causing cancer.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECSNumber DA8340700

RUO – Research Use Only