Learn More
Thermo Scientific™ SickleScreen™ Assay Kit
Screen for sickle cell disease and sickle cell trait with this assay kit.
Supplier:  Thermo Scientific™ 100250
 View more versions of this product
Catalog No. CMS-251-050
Only null left
Description
Description
Screen for sickle cell disease and sickle cell trait with the Fisherbrand™ Pacific Hemostasis™ SickleScreen™ Assay Kit.
Thermo Scientific SickleScreen products use a modified Nalbandian procedure that provides fast and accurate results for screening and detecting hereditary sickle cell disease and sickle cell trait. When sickle cell patients are in crisis, their Hb S becomes sickle or cresent shaped and can no longer carry oxygen. The abnormal cells often break apart and can cause damage to the spleen, kidneys, bone, eyes, lungs and central nervous system.
  • The procedure is based on the differential solubility of Hemoglobin S (Hb S) and Hemoglobin A (Hb A) in concentrated phosphate solution.
  • The 120-test kit contains phosphate buffer (4 x 125mL) and four sodium hydrosulfite vials.
  • The 30-test kit contains a bottle of phosphate buffer (125mL) with a dispensing cap and 30 pre-filled reaction vials
  • Positive and negative controls are for Hb A and Hb S.
Ordering Alerts:
The control kit (22-251-051) must be ordered separately.
Specifications
Specifications
SickleScreen Assay Set
Prefilled vials with sodium hydrosulfite powder, phosphate buffer 1 x 125 mL with dispensing cap.
30-Determination Kit
Product Suggestions
Product Suggestions