missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pacific Hemostasis™ Prothrombin Time (PT) Reagents

Catalog No. CMS176065
Change view
Click to view available options
Quantity:
10 x 4 mL
10 x 10 mL
Type:
Prothrombin Time (PT)
Thromboplastin-DS
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
CMS176065 10 x 4 mL Prothrombin Time (PT)
292273 10 x 4 mL Thromboplastin-DS
292274 10 x 10 mL Thromboplastin-DS
176099 10 x 10 mL Prothrombin Time (PT)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. CMS176065 Supplier Thermo Scientific™ Supplier No. 10 0356

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Detect acquired or congenital disorders affecting the extrinsic and common pathways.

Detect acquired or congenital disorders affecting the extrinsic and common pathways with Thermo Scientific™ Pacific Hemostasis™ Prothrombin Time (PT) Reagents. Widely accepted as a means of monitoring patients on oral anticoagulant therapy such as Warfarin/Coumadin, reagents are manufactured with up to three years extended shelf life and have excellent lot-to-lot consistency.

Features:
  • Compatible with automated, semi-automated or manual systems
  • Reconstitution stability: 7 days at 2° to 8°C (36° to 46°F)
  • Dried rabbit brain thromboplastin

Thromboplastin-D with CaCl2 (Product No. 10-0356, 10-0357):

  • Target ISI value of 1.7 to 2.2
  • Expected normal values of 12 to 15 seconds

Thromboplastin-DS (Product No. 10-0362, 10-0354):

  • Target ISI value of 1.0 to 1.3
  • Expected normal values of 12 to 15 seconds

Specifications

Type Prothrombin Time (PT)
Quantity 10 x 4 mL