missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pacific Hemostasis™ Imidazole Buffered Saline

Catalog No. p-4528068
Change view
Click to view available options
Type:
Fibrinogen Assay
Fibrinogen Reference
Quantity:
100 Determinations
10 x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity
CMS176305 Fibrinogen Reference 10 x 1 mL
22176271 Fibrinogen Assay 100 Determinations
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CMS176305 Supplier Thermo Scientific™ Supplier No. 10 0602
Only null left

Fisherbrand™ Pacific Hemostasis Fibrinogen/Fibrin Analysis is a complete line of assays used to screen and quantify plasma fibrinogen based on the Clauss method.

Screen and quantify plasma fibrinogen based on the Clauss method with Thermo Scientific™ Pacific Hemostasis™ Fibrinogen/Fibrin Analysis.

Fibrinogen Assay Set:
  • Quantifies plasma fibrinogen levels
  • Detects acquired and congenital hemorrhagic disorders
  • Components available separately
Thrombin Time Assay:
  • Provides qualitative determination
  • Useful in screening for fibrinogen disorders by measuring the formation of the fibrin clot
  • Reconstituted stability: 8 hours at 2° to 8°C

Ordering Information:

Products can be purchased as a set or individually.

Specifications

Content And Storage 2°C to 8°C
Type Fibrinogen Reference
Quantity 10 x 1 mL