Learn More
Thermo Scientific™ Pacific Hemostasis™ Coagulation Controls
Provide accurate, reproducible confirmation of coagulation test system performance with these coagulation controls.
Supplier: Thermo Scientific™ 100604
Description
Provide accurate, reproducible confirmation of coagulation test system performance with Thermo Scientific™ Pacific Hemostasis™ Coagulation Controls. Controls can be run on most coagulation systems and are formulated to reflect expected patient results.
Coagulation and Heparin control plasmas are prepared from fresh pools of human plasma.
Coagulation Control Plasmas:
- Normal control (Level 1) produces normal PT and aPTT times
- Abnormal controls monitor therapeutic (Level 2) and highly elevated (Level 3) PT/aPTT times
Heparin Control Plasmas:
- Simulate low (Level 1) and elevated (Level 2) plasma levels of Heparin
- Use to monitor and evaluate patients on Heparin therapy
Spécifications
| Fibrinogen Low Control | |
| 10 x 1 mL |