Learn More
Thermo Scientific™ Pacific Hemostasis™ Coagulation Controls
Provide accurate, reproducible confirmation of coagulation test system performance with these coagulation controls.
Supplier:  Thermo Scientific™ 10 0597
 View more versions of this product
Catalog No. CMS176230
Only null left
Description
Description
Provide accurate, reproducible confirmation of coagulation test system performance with Thermo Scientific™ Pacific Hemostasis™ Coagulation Controls. Controls can be run on most coagulation systems and are formulated to reflect expected patient results.
Coagulation and Heparin control plasmas are prepared from fresh pools of human plasma.
Coagulation Control Plasmas:
  • Normal control (Level 1) produces normal PT and aPTT times
  • Abnormal controls monitor therapeutic (Level 2) and highly elevated (Level 3) PT/aPTT times 
Heparin Control Plasmas:
  • Simulate low (Level 1) and elevated (Level 2) plasma levels of Heparin
  • Use to monitor and evaluate patients on Heparin therapy
Specifications
Chemical Identifiers
10 x 1 mL
Specifications
Coagulation Control Level 3
10 x 1 mL
Product Suggestions
Product Suggestions