missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pacific Hemostasis™ Activated Partial Thromboplastin Time (APTT) Reagents

Catalog No. CMS285545
Change view
Click to view available options
Description:
aPTT-XL
APTT-XL (Ellagic Acid Activator)
CaCl2, 0.02M
Quantity:
10 x 4 mL
10 x 10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Quantity
CMS285545 CaCl2, 0.02M 10 x 10 mL
CMS022929 APTT-XL (Ellagic Acid Activator) 10 x 4 mL
CMS022927 aPTT-XL 10 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CMS285545 Supplier Thermo Scientific™ Supplier No. 10 0304

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fisherbrand™ Pacific Hemostasis Activated Partial Thromboplastin Time (aPTT) Reagents are manufactured with up to two years extended shelf life.

Identify acquired or congenital disorders to the intrinsic and common pathways with Thermo Scientific™ Pacific Hemostasis™ Activated Partial Thromboplastin Time (APTT) Reagents. Widely accepted as a means of monitoring patients on oral anticoagulant therapy such as Heparin, reagents are manufactured with up to two years extended shelf life and have excellent lot-to-lot consistency.

Pacific Hemostasis Activated Partial Thromboplastin Time reagents (APTT) are used to identify acquired or congenital disorders to the intrinsic and common pathways. APTT testing is widely accepted as a means of monitoring patients on oral anticoagulant therapy such as heparin.
  • 10mL vials can complete ∽1000 full volume, single determinations; 4mL vials can complete ∽400 full volume, single determinations
  • Open vial stability: 30 days at 2° to 8°C

aPTT-XL (Ellagic Acid Activator) (Product No. 10-0402)

  • Superior factor sensitivity and responsiveness
  • Increased sensitivity to heparin and lupus-like inhibitors

KONTACT

™(Particulate Activator) (Product No. 01-0402)

  • Higher sensitivity to Factor VIII
  • Average Normal Values: 26 - 33 Seconds

Specifications

Description CaCl2, 0.02M
Type Activated Partial Thromboplastin Time (APTT)
Quantity 10 x 10 mL