missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pacific Hemostasis™ Activated Partial Thromboplastin Time (APTT) Reagents
Description
Identify acquired or congenital disorders to the intrinsic and common pathways with Thermo Scientific™ Pacific Hemostasis™ Activated Partial Thromboplastin Time (APTT) Reagents. Widely accepted as a means of monitoring patients on oral anticoagulant therapy such as Heparin, reagents are manufactured with up to two years extended shelf life and have excellent lot-to-lot consistency.
Features:
- 10mL vials can complete ∽1000 full volume, single determinations; 4mL vials can complete ∽400 full volume, single determinations
- Open vial stability: 30 days at 2° to 8°C
aPTT-XL (Ellagic Acid Activator) (Product No. 10-0402):
- Superior factor sensitivity and responsiveness
- Increased sensitivity to heparin and lupus-like inhibitors
KONTACT™ (Particulate Activator) (Product No. 01-0402):
- Higher sensitivity to Factor VIII
- Average Normal Values: 26 - 33 Seconds
Specifications
Specifications
| Description | APTT-XL (Ellagic Acid Activator) |
| Type | Activated Partial Thromboplastin Time (APTT) |
| Quantity | 10 x 4 mL |