Learn More
SignalChem Biotech RSK2, Active 500ug
Catalog No. 502573238
Encompass
Recombinant full-length human RSK2 was expressed by baculovirus in Sf9 insect cells using a N-terminal GST tag.
Catalog No. 502573238 Supplier SignalChem Biotech Supplier No. R1710G500
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant full-length human RSK2 was expressed by baculovirus in Sf9 insect cells using a N-terminal GST tag.