Learn More
Enzo Life Sciences p62 (human) polyclonal antibody (100 l) DFS Item
Supplier:  Enzo BMLPW98600100
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 387-436 human p62. UniProt: Q13501. Source: Purified from rabbit serum. Species Reactivity: Human. Applications: ELISA, IF, IHC, WB. Recommended dilutions: Immunohistochemistry (1:1,000). Western Blot (1:1,000). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 0.09% sodium azide. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20C.
Catalog No. 501031794
Only null left
Explore available promotions