Learn More
SignalChem Biotech p33ING1 Protein 1000ug
Catalog No. 502574618
Encompass
Recombinant full-length human p33ING1 was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
Catalog No. 502574618 Supplier SignalChem Biotech Supplier No. I3230H1000
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant full-length human p33ING1 was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.