Learn More
Promega P450-Glo™ CYP3A4 Assay with Luciferin-IPA
The P450-Glo CYP3A4 Assay System provides a homogeneous, luminescent method for measuring CYP3A4 activity and the screening system contains a complete set of reagents for performing the assay.
Supplier:  Promega V9001
Encompass
 View more versions of this product
Catalog No. PRV9001
Only null left
Includes: Luciferin Detection Reagent; P450-Glo Buffer; 5mM Luciferin-CEE, -ME, -H, -BE, -PFBE, -PPXE, or IPA
Description
Description
  • Luminescent Assays and Screening Systems for Measuring CYP3A4 Activity
  • Screening systems include complete set of reagents
  • Luminescent format eliminates need for time-consuming analyses
  • Broad dynamic range and low background
Specifications
Chemical Identifiers
-30°C to -10°C
Homogeneous, Luminescence
For measuring cytochrome P450 activity
P450-Glo CYP3A4 Assay with Luciferin-IPA
Specifications
10 mL
Homogeneous, Luminescence
1 x 1 Each Luciferin detection reagent, 1 x 10mL Reconstitution Buffer with esterase, 1 x 15μL Luciferin-IPA
-30°C to -10°C
For measuring cytochrome P450 activity
P450-Glo CYP3A4 Assay with Luciferin-IPA
Product Suggestions
Product Suggestions