missing translation for 'onlineSavingsMsg'
Learn More

Promega P450-Glo CYP2B6 Assay, 50ml

Catalog No. PRV8322
Encompass
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV8322 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV8322 Supplier Promega Supplier No. V8322

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The P450-Glo Screening Systems provide a complete set of reagents for performing luminescent cytochrome P450 assays

  • A Luminescent Method and Screening System for Measuring CYP2B6 Activity
  • Screening system includes complete set of reagents
  • Luminescent format eliminates need for time−consuming analyses
  • Broad dynamic range and low background

Specifications

Quantity 50 mL
Content And Storage -30°C to -10°C