missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega P450-Glo CYP2B6 Assay, 10ml
Description
- A Luminescent Method and Screening System for Measuring CYP2B6 Activity
- Screening system includes complete set of reagents
- Luminescent format eliminates need for time−consuming analyses
- Broad dynamic range and low background
Specifications
Specifications
| Quantity | 10 mL |
| Content And Storage | -30°C to -10°C |