UserName
Recombinant full-length human CDC7 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.