missing translation for 'onlineSavingsMsg'
Learn More

p-Xyloquinone 98.0+%, TCI America™

Catalog No. D06865G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D06865G 5 g
D06861G 1 g
D068625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D06865G Supplier TCI America Supplier No. D06865G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 137-18-8
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00041737
InChI Key MYKLQMNSFPAPLZ-UHFFFAOYSA-N
Synonym 2,5-dimethyl-1,4-benzoquinone, phlorone, p-xyloquinone, 2,5-dimethyl-p-benzoquinone, 2,5-xyloquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-dimethyl, 2,5-dimethylquinone, floron, 3,6-dimethyl-p-benzoquinone, floron czech
PubChem CID 8718
IUPAC Name 2,5-dimethylcyclohexa-2,5-diene-1,4-dione
SMILES CC1=CC(=O)C(=CC1=O)C

Specifications

Melting Point 125°C
Quantity 5 g
Formula Weight 136.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material p-Xyloquinone
RTECSNumber DK4800000
TSCA Yes