missing translation for 'onlineSavingsMsg'
Learn More

p-Xylene 99.0+%, TCI America™

Catalog No. X0014500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X0014500ML 500 mL
X001425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. X0014500ML Supplier TCI America Supplier No. X0014500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 106-42-3
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008556
InChI Key URLKBWYHVLBVBO-UHFFFAOYSA-N
Synonym 1,4-Dimethylbenzene
PubChem CID 7809
ChEBI CHEBI:27417
IUPAC Name 1,4-xylene
SMILES CC1=CC=C(C)C=C1

Specifications

Boiling Point 139°C
Physical Form Clear Liquid at 20°C
Color Colorless
Quantity 500 mL
UN Number 1307
Formula Weight 106.17
Percent Purity ≥99.0% (GC)
Chemical Name or Material p-Xylene
EINECSNumber (3)-0003&(3)-0060
RTECSNumber ZE2625000
TSCA Yes