missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ PVac™ Portable Vacuum System

Catalog No. 03391217
Change view
Click to view available options
Flow Rate:
4 L/min.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate
03391217 4 L/min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03391217 Supplier Cole-Parmer™ Supplier No. 0439707

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

filter, tubing, battery, power supply and HandEvac hand operator with single channel adapter

Construct your own waste aspiration system—no other vacuum source needed!

  • Take into the field and run off the rechargeable battery to filter samples on site
  • Compact, portable liquid waste disposal device accommodates various collection vessels
  • Operate in the lab on the universal power adapter—power supply also acts as battery charger
  • Hydrophobic filter protects pump from over-filling
  • 1 year warranty
Combine this oil-free, diaphragm vacuum pump with a collection bottle to create a portable waste aspiration system and rapidly remove supernatant from a variety of cell culture or multiwell plates and dishes. The vacuum system can also be used as a stand-alone pump for vacuum-assisted filtration applications like bottletop and vacuum manifold filtration.

Compact size makes the vacuum system easily portable and allows for placement in an individual workstation or under a hood. A 0.45-μm hydrophobic, in-line filter protects from overfilling or fluid aspiration into the pump—ideal for filtering hazardous samples too dangerous for house lines, and as the vacuum source for waste aspiration systems. Couple with any glass or plastic collection bottle or flask—like the Mini-Vac waste collection bottle—and empty a 2-liter bottle of waste in 40 seconds!

The HandE-Vac handheld aspirator (included) lets you control the vacuum pressure and speed by manually adjusting aperture button. An adapter for a single channel pipette is included; order different adapters for aspiration of multiwell plates and Pasteur pipettes. *Collection bottle used with the P-Vac vacuum system should not exceed 2 liters.

Certifications

UL, CE, and CSA approved

Specifications

Max. Vacuum -250 to -500 mbar
Control Analog dial
Includes Filter, tubing, battery (2+ hours of continuous use), power supply and HandE-Vac hand operator with single channel adapter
Certifications/Compliance UL, CSA and CE
Electrical Requirements 100/240 VAC 50/60 Hz
Description P-Vac system includes filter, tubing, battery, power supply and HandEvac hand operator with single channel adapter
Model PV000
Plug Type Four regional styles included with universal power supply
Weight (English) 2 lb.
Weight (Metric) 910 g
Flow Rate 4 L/min.
Frequency 50/60 Hz
Pump Type Oilless Diaphragm Pump
Applications Portable vacuum source for filtration and waste aspiration
Maximum Pressure -500 mbar
For Use With (Equipment) M-Vac, M-Vac Jr., HandE-Vac, and other vacuum-approved systems
Voltage 100/240 VAC
Show More Show Less