missing translation for 'onlineSavingsMsg'
Learn More

p-Tolyltrichlorosilane, 97%, Thermo Scientific Chemicals

Catalog No. p-7025974
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2172806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2172806 Supplier Thermo Scientific Chemicals Supplier No. B2172806

It can be used to produce 3-(p-Methylphenyl)cyclohexanon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used to produce 3-(p-Methylphenyl)cyclohexanon.

Solubility
Reacts with water.

Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 701-35-9
Molecular Formula C7H7Cl3Si
Molecular Weight (g/mol) 225.568
MDL Number MFCD00045131
InChI Key WOMUGKOOLXQCTQ-UHFFFAOYSA-N
Synonym p-tolyltrichlorosilane, tolyltrichlorosilane, trichloro-p-tolylsilane, silane, trichloro 4-methylphenyl, trichloro 4-methylphenyl silane, trichloro p-tolyl silane, silane, trichloro-p-tolyl, benzene, 1-methyl-4-trichlorosilyl, 4-tolyltrichlorosilane, p-tolylsilyl trichloride
PubChem CID 69697
IUPAC Name trichloro-(4-methylphenyl)silane
SMILES CC1=CC=C(C=C1)[Si](Cl)(Cl)Cl

Specifications

Boiling Point 218°C to 220°C
Quantity 5 g
UN Number UN2987
Beilstein 1866557
Flash Point 98°C (208°F)
Solubility Information Reacts with water.
Formula Weight 225.58
Percent Purity 97%
Odor Irritating
Refractive Index 1.524
Sensitivity Moisture sensitive
Density 1.28
Chemical Name or Material p-Tolyltrichlorosilane
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.
EINECSNumber 211-854-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only