missing translation for 'onlineSavingsMsg'
Learn More

p-Tolylacetylene, 97%

Catalog No. AC392560010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC392560010 1 g Glass bottle
AC392560050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC392560010 Supplier Thermo Scientific Chemicals Supplier No. 392560010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 766-97-2 | C9H8 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 766-97-2
Molecular Formula C9H8
Molecular Weight (g/mol) 116.16
MDL Number MFCD00008571
InChI Key KSZVOXHGCKKOLL-UHFFFAOYSA-N
Synonym 4-ethynyltoluene, p-tolylacetylene, 4-methylphenylacetylene, p-ethynyltoluene, p-methylphenylacetylene, benzene, 1-ethynyl-4-methyl, toluene, p-ethynyl, benzene, 1-ethynyl-4-methyl-9ci, 4'-methylphenyl acetylene, 1-ethynyl-4-methyl-benzene
PubChem CID 13018
IUPAC Name 1-ethynyl-4-methylbenzene
SMILES CC1=CC=C(C=C1)C#C

Specifications

Color Yellow
Density 0.9160g/mL
Boiling Point 168.0°C to 170.0°C
Flash Point 49°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5460 to 1.5480 (20°C, 589nm)
Linear Formula CH3C6H4C≡CH
Quantity 1 g
Specific Gravity 0.916
Solubility Information Solubility in water: insoluble
Formula Weight 116.16
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material p-Tolylacetylene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only