missing translation for 'onlineSavingsMsg'
Learn More

p-Tolylacetic Acid 98.0+%, TCI America™

Catalog No. p-7136479
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T088225G 25 g
T0882250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T088225G Supplier TCI America Supplier No. T088225G
Only null left

Chemical Identifiers

CAS 622-47-9
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00004353
InChI Key GXXXUZIRGXYDFP-UHFFFAOYSA-N
Synonym p-tolylacetic acid, 4-methylphenylacetic acid, 2-p-tolyl acetic acid, 2-4-methylphenyl acetic acid, p-methylphenylacetic acid, 4-tolylacetic acid, 4-methylphenyl acetic acid, benzeneacetic acid, 4-methyl, acetic acid, p-tolyl, 2-p-tolylacetic acid
PubChem CID 248474
IUPAC Name 2-(4-methylphenyl)acetic acid
SMILES CC1=CC=C(C=C1)CC(=O)O

Specifications

Melting Point 92°C
Color White-Yellow
Boiling Point 267°C
Quantity 25 g
Formula Weight 150.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material p-Tolylacetic Acid
RTECSNumber AJ7569000
TSCA No