missing translation for 'onlineSavingsMsg'
Learn More

p-Tolyl Isocyanate 98.0+%, TCI America™

Catalog No. p-7136220
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T032125G 25 g
T0321100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T032125G Supplier TCI America Supplier No. T032125G
Only null left

Chemical Identifiers

CAS 622-58-2
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00002029
InChI Key MGYGFNQQGAQEON-UHFFFAOYSA-N
Synonym p-tolyl isocyanate, 4-tolyl isocyanate, 4-methylphenyl isocyanate, p-isocyanatotoluene, 4-isocyanatotoluene, p-toluene isocyanate, p-methylphenyl isocyanate, benzene, 1-isocyanato-4-methyl, p-tolylisocyanate, 1-isocyanato-4-methyl-benzene
PubChem CID 69325
ChEBI CHEBI:53732
IUPAC Name 1-isocyanato-4-methylbenzene
SMILES CC1=CC=C(C=C1)N=C=O

Specifications

Color Yellow
Boiling Point 74°C
Quantity 25 g
UN Number 2206
Formula Weight 133.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material p-Tolyl Isocyanate
EINECSNumber (3)-3238
TSCA Yes
Recommended Storage Refrigerator