missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Toluquinone 98.0+%, TCI America™
$127.07 - $1226.33
Chemical Identifiers
| CAS | 553-97-9 |
|---|---|
| Molecular Formula | C7H6O2 |
| Molecular Weight (g/mol) | 122.12 |
| MDL Number | MFCD00001603 |
| InChI Key | VTWDKFNVVLAELH-UHFFFAOYSA-N |
| Synonym | p-toluquinone, methyl-p-benzoquinone, 2-methyl-1,4-benzoquinone, toluquinone, methyl-1,4-benzoquinone, 2-methyl-p-benzoquinone, tolylquinone, 2-methylbenzoquinone, 2-methylquinone, methylbenzoquinone |
| PubChem CID | 11122 |
| IUPAC Name | 2-methylcyclohexa-2,5-diene-1,4-dione |
| SMILES | CC1=CC(=O)C=CC1=O |
Chemical Identifiers
| 553-97-9 | |
| 122.12 | |
| VTWDKFNVVLAELH-UHFFFAOYSA-N | |
| 11122 | |
| CC1=CC(=O)C=CC1=O |
| C7H6O2 | |
| MFCD00001603 | |
| p-toluquinone, methyl-p-benzoquinone, 2-methyl-1,4-benzoquinone, toluquinone, methyl-1,4-benzoquinone, 2-methyl-p-benzoquinone, tolylquinone, 2-methylbenzoquinone, 2-methylquinone, methylbenzoquinone | |
| 2-methylcyclohexa-2,5-diene-1,4-dione |
Specifications
| 553-97-9 | |
| Yellow | |
| MFCD00001603 | |
| p-toluquinone, methyl-p-benzoquinone, 2-methyl-1,4-benzoquinone, toluquinone, methyl-1,4-benzoquinone, 2-methyl-p-benzoquinone, tolylquinone, 2-methylbenzoquinone, 2-methylquinone, methylbenzoquinone | |
| CC1=CC(=O)C=CC1=O | |
| 122.12 | |
| 122.12 | |
| Crystalline Powder |
| 68°C | |
| C7H6O2 | |
| 25 g | |
| VTWDKFNVVLAELH-UHFFFAOYSA-N | |
| 2-methylcyclohexa-2,5-diene-1,4-dione | |
| 11122 | |
| ≥98.0% (GC,T) | |
| p-Toluquinone |
Safety and Handling
EINECSNumber : (3)-1002
RTECSNumber : DK6300000
TSCA : Yes