missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Toluquinone 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 68°C |
| Color | Yellow |
| Quantity | 25 g |
| Formula Weight | 122.12 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | p-Toluquinone |
Safety and Handling
| EINECSNumber | (3)-1002 |
| RTECSNumber | DK6300000 |
| TSCA | Yes |