missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Toluoyl Chloride 98.0+%, TCI America™
Supplier: TCI America T0311100G
Specifications
| p-Toluoyl Chloride | |
| Yellow | |
| C8H7ClO | |
| 100 g | |
| p-toluoyl chloride, benzoyl chloride, 4-methyl, p-methylbenzoyl chloride, 4-toluoyl chloride, p-toluyl chloride, p-toluic acid chloride, 4-methylbenzoic acid chloride, 4-methylbenzoylchloride, 4-methyl-benzoyl chloride, unii-092e36pwl4 | |
| CC1=CC=C(C=C1)C(=O)Cl | |
| 154.593 | |
| 154.59 | |
| Liquid |
| 874-60-2 | |
| 216°C | |
| MFCD00000696 | |
| 3265 | |
| NQUVCRCCRXRJCK-UHFFFAOYSA-N | |
| 4-methylbenzoyl chloride | |
| 13405 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 874-60-2 | |
| 154.593 | |
| NQUVCRCCRXRJCK-UHFFFAOYSA-N | |
| 13405 | |
| CC1=CC=C(C=C1)C(=O)Cl |
| C8H7ClO | |
| MFCD00000696 | |
| p-toluoyl chloride, benzoyl chloride, 4-methyl, p-methylbenzoyl chloride, 4-toluoyl chloride, p-toluyl chloride, p-toluic acid chloride, 4-methylbenzoic acid chloride, 4-methylbenzoylchloride, 4-methyl-benzoyl chloride, unii-092e36pwl4 | |
| 4-methylbenzoyl chloride |
Safety and Handling
EINECSNumber : (3)-3707
RTECSNumber : XV1660000
TSCA : Yes