Learn More
p-Toluoyl chloride, 99%
CAS: 874-60-2 | C8H7ClO | 154.593 g/mol
Supplier: Thermo Scientific Chemicals A155810E
Description
p-Toluoyl chloride was used to prepare antigen, Tolyl-amido-human serum albumin. It is used as a reagent to synthesize (+)-Yashabushitriol, derivatives of Desloratadine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsp-Toluoyl chloride was used to prepare antigen, Tolyl-amido-human serum albumin. It is used as a reagent to synthesize (+)-Yashabushitriol, derivatives of Desloratadine.
Solubility
Reacts with water alcohol.
Notes
Store at room temperature. Incompatible with oxidizing agent, acids, bases, active metals, alcohols, amines.
Specifications
| p-Toluoyl chloride | |
| -2°C | |
| 225°C to 227°C | |
| 99% | |
| C8H7ClO | |
| MFCD00000696 | |
| UN3265 | |
| Moisture sensitive | |
| Reacts with water alcohol. | |
| CC1=CC=C(C=C1)C(=O)Cl | |
| 154.593 | |
| 154.6 | |
| Liquid |
| 874-60-2 | |
| 1.169 | |
| 82°C (179°F) | |
| Pungent | |
| 1.553 | |
| 2500 g | |
| 471492 | |
| p-toluoyl chloride, benzoyl chloride, 4-methyl, p-methylbenzoyl chloride, 4-toluoyl chloride, p-toluyl chloride, p-toluic acid chloride, 4-methylbenzoic acid chloride, 4-methylbenzoylchloride, 4-methyl-benzoyl chloride, unii-092e36pwl4 | |
| NQUVCRCCRXRJCK-UHFFFAOYSA-N | |
| 4-methylbenzoyl chloride | |
| 13405 | |
| 99% |
Chemical Identifiers
| 874-60-2 | |
| 154.593 | |
| NQUVCRCCRXRJCK-UHFFFAOYSA-N | |
| 13405 | |
| CC1=CC=C(C=C1)C(=O)Cl |
| C8H7ClO | |
| MFCD00000696 | |
| p-toluoyl chloride, benzoyl chloride, 4-methyl, p-methylbenzoyl chloride, 4-toluoyl chloride, p-toluyl chloride, p-toluic acid chloride, 4-methylbenzoic acid chloride, 4-methylbenzoylchloride, 4-methyl-benzoyl chloride, unii-092e36pwl4 | |
| 4-methylbenzoyl chloride |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 212-864-8
RTECSNumber : XV1660000
TSCA : Yes
storageNote1 : Moisture sensitive
Recommended Storage : Ambient temperatures
RUO – Research Use Only