missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich P-Toluenesulfonyl Chloride
Description
P-Toluenesulfonyl Chloride
Specifications
Specifications
| CAS | 98-59-9 |
| Melting Point | 65°C to 69°C |
| Boiling Point | 134°C (10 mmHg, lit.) |
| Molecular Formula | C7H7ClO2S |
| Linear Formula | CH3C6H4SO2Cl |
| MDL Number | MFCD00007450 |
| Quantity | 500 g |
| Synonym | TsCl; Tosyl chloride |
| Molecular Weight (g/mol) | 190.65 g/mol |