missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonic anhydride, 95%, Thermo Scientific™

Chemical Identifiers

CAS 4124-41-8
Molecular Formula C14H14O5S2
Molecular Weight (g/mol) 326.39
MDL Number MFCD00008548
InChI Key PDVFSPNIEOYOQL-UHFFFAOYSA-N
Synonym p-toluenesulfonic anhydride, 4-methylbenzenesulfonic anhydride, 4-toluenesulfonic anhydride, p-toluenesulfonicanhydride, p-toluene sulfonic anhydride, toluene-p-sulphonic anhydride, 4-methylphenyl sulfonyl 4-methylbenzenesulfonate, p-toluenesulphonic anhydride, toluene-4-sulfonic anhydride, benzenesulfonic acid, 4-methyl-, anhydride
PubChem CID 77773
IUPAC Name (4-methylphenyl)sulfonyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OS(=O)(=O)C2=CC=C(C=C2)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC367070050
SDS View Documents
Thermo Scientific Chemicals
367070050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC367070250
SDS View Documents
Thermo Scientific Chemicals
367070250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC367071000
SDS View Documents
Thermo Scientific Chemicals
367071000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

4124-41-8
326.39
PDVFSPNIEOYOQL-UHFFFAOYSA-N
77773
CC1=CC=C(C=C1)S(=O)(=O)OS(=O)(=O)C2=CC=C(C=C2)C
C14H14O5S2
MFCD00008548
p-toluenesulfonic anhydride, 4-methylbenzenesulfonic anhydride, 4-toluenesulfonic anhydride, p-toluenesulfonicanhydride, p-toluene sulfonic anhydride, toluene-p-sulphonic anhydride, 4-methylphenyl sulfonyl 4-methylbenzenesulfonate, p-toluenesulphonic anhydride, toluene-4-sulfonic anhydride, benzenesulfonic acid, 4-methyl-, anhydride
(4-methylphenyl)sulfonyl 4-methylbenzenesulfonate

Specifications

4124-41-8
95%
C14H14O5S2
5g
p-toluenesulfonic anhydride, 4-methylbenzenesulfonic anhydride, 4-toluenesulfonic anhydride, p-toluenesulfonicanhydride, p-toluene sulfonic anhydride, toluene-p-sulphonic anhydride, 4-methylphenyl sulfonyl 4-methylbenzenesulfonate, p-toluenesulphonic anhydride, toluene-4-sulfonic anhydride, benzenesulfonic acid, 4-methyl-, anhydride
CC1=CC=C(C=C1)S(=O)(=O)OS(=O)(=O)C2=CC=C(C=C2)C
326.39
326.39
p-Toluenesulfonic anhydride
Authentic
Glass bottle
MFCD00008548
11, IV, 376
PDVFSPNIEOYOQL-UHFFFAOYSA-N
(4-methylphenyl)sulfonyl 4-methylbenzenesulfonate
77773
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 223-926-9

SDS
Documents

Documents

RUO – Research Use Only