missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonic acid monohydrate, 97%, Thermo Scientific Chemicals

Catalog No. AAA1411922
Change view
Click to view available options
Quantity:
100 g
500 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1411922 100 g
AAA1411936 500 g
AAA141190I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1411922 Supplier Thermo Scientific Chemicals Supplier No. A1411922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

p-Toluenesulfonic acid monohydrate is used as a catalyst in the synthesis of resveratrol, in oxane derivatives as an antimalarial agent, as substituted piperidine and unsymmetrical benzyl. It acts as a catalyst in the preparation of 1,3,5-trisubstituted pyrazoles derivatives, selenated ketene dithioacetals, triazoloquinazolinone and benzimidazoquinazolinone derivatives. It also serves as an intermediate in the esterification and in reductive amination reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Toluenesulfonic acid monohydrate is used as a catalyst in the synthesis of resveratrol, in oxane derivatives as an antimalarial agent, as substituted piperidine and unsymmetrical benzyl. It acts as a catalyst in the preparation of 1,3,5-trisubstituted pyrazoles derivatives, selenated ketene dithioacetals, triazoloquinazolinone and benzimidazoquinazolinone derivatives. It also serves as an intermediate in the esterification and in reductive amination reactions.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 6192-52-5
Molecular Formula C7H10O4S
Molecular Weight (g/mol) 190.213
MDL Number MFCD00142137
InChI Key KJIFKLIQANRMOU-UHFFFAOYSA-N
Synonym p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate
PubChem CID 521998
IUPAC Name 4-methylbenzenesulfonic acid;hydrate
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.O

Specifications

Melting Point 103°C to 106°C (anhydrous)
Boiling Point 140°C (20mmHg)
UN Number UN2585
Beilstein 3568023
Merck Index 14,9533
Formula Weight 190.22 (172.20 Anhydrous)
Percent Purity 98%
Density 1.24
Quantity 100 g
Flash Point 150°C (302°F)
Odor Mild
Sensitivity Hygroscopic
Chemical Name or Material p-Toluenesulfonic acid monohydrate
Show More Show Less
Hazard Category H314-H335-H373
Hazard Statement GHS H Statement
H314-H335-H319
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, SOLID
EINECSNumber 203-180-0
RTECSNumber DB7164000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only